ContactUsLinkComponent
Merlin Microseal™ Septa for Shimadzu GCs, Restek
Catalogue # 10855-840
Fournisseur:  Restek
undefined
Merlin Microseal™ Septa for Shimadzu GCs, Restek
Catalogue # 10855-840
Fournisseur:  Restek
Supplier Number:  22972
Spécifications
  • Description:
    General purpose kit for Shimadzu GCs (3 - 100 psi)
  • Cat. no.:
    10855-840
  • Supplier No.:
    22972
Spécifications
À propos de cet article
A Merlin Microseal septum provides several distinct advantages: elimination of septum coring, long lifetime, and consistently low needle-insertion force.
  • Compatible with Shimadzu models GC-2010 and GC-2025
  • 450 °C maximum injection port temperature
  • For use with 23-gauge (0.63 mm, 0.025") needle or probe with blunt, truncated conical tip
The Microseal septum incorporates two separate sealing mechanisms; these sliding seals prevent septum coring and the associated accumulation of septum crumbs in the injection port liner. Installation is simple, requiring no modification of the injection port.
Delivery: Includes Nut, Adapter, O-Ring, and 2 General-Purpose (#410) Microseals.