Heater Block for Agilent Injectors, Restek
Catalog # 10856-168
Supplier: Restek
Specifications
- Description:Heater Block
- Environmentally Preferable:
- Cat. no.:10856-168
- Supplier No.:27199
Specifications
About this item
For use with Agilent 6850, 6890, and 7890 GC split/splitless injectors.
- Meets or exceeds original manufacturer's performance