Ferrules for SRI & Buck Scientific GCs, Fit 0.25/0.32 mm int.Ø MXT® Columns, Restek
Fournisseur: Restek
Certificats
À propos de cet article
Fits columns with internal diameters of 0.25 mm or 0.32 mm
- Durable, leak-tight
- Preconditioned to eliminate out-gassing
- Seal with minimal torque, reusable, and preferred for vacuum and high-pressure uses
- Recommended for mass spec transfer lines
For use with SRI and Buck Scientific GCs.
Frequently Bought Together