Ferrules for SRI & Buck Scientific GCs, Fit 0.25/0.32 mm int.Ø Fused Silica Columns, Restek
Supplier: Restek
Certificates
About this item
Fits columns with internal diameters of 0.25 mm or 0.32 mm
- Durable, leak-tight
- Preconditioned to eliminate out-gassing
- Seal with minimal torque, reusable, and preferred for vacuum and high-pressure uses
- Recommended for mass spec transfer lines
For use with SRI and Buck Scientific GCs.
Frequently Bought Together