Compact Ferrules for Agilent GCs, Fit 0.25/0.32 mm int.Ø MXT® Columns, Restek
Fournisseur: Restek
Certificats
À propos de cet article
Compact ferrules are ideal for use with Agilent GCs
- Preconditioned to eliminate outgassing
- High-purity, high-density graphite or Vespel®/graphite material
- Stable to 400°C
Ferrules feature internal diameters of 0.5 mm, compatible with columns 0.32 mm in diameter.
Frequently Bought Together