ContactUsLinkComponent
Parker Balston® Nitrogen Gas Generators for LC-MS, Restek
Catalogue # 10058-726
Fournisseur:  Restek
undefined
Parker Balston® Nitrogen Gas Generators for LC-MS, Restek
Catalogue # 10058-726
Fournisseur:  Restek
Supplier Number:  22131
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Nitrogen Generator for LC-MS, N2-22 Model
  • Cat. no.:
    10058-726
  • Supplier No.:
    22131
Spécifications
À propos de cet article
This nitrogen gas generators turn compressed air into ultra-pure nitrogen (up to 99.5%).
  • Flows from 1 to 44 L/min
  • Models N2-04, N2-14, N2-22, and N2-35 require no electricity
  • Safe and reliable
  • Low maintenance
  • Maintenance kits include replacement filters
Informations sur les accessoires: For use with Parker Balston® model FID-1000 and FID-2500 gas stations and Filters should be changed every year, resin and desiccant cartridges can be replaced every 2 years.