Single Taper Inlet Liner with Wool for Bruker/Varian GCs equipped with 1177 inlets, Restek
Fournisseur: Restek
Certificats
À propos de cet article
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
- Consistent quality with exceptional batch-to-batch uniformity
- Low to zero bleed or background contamination
- Internal diameter of 4 mm
Superior coverage is delivered even with highly active compounds.
Frequently Bought Together