Double Taper Inlet Liner for Thermo TRACE, 8000 Series and Focus GCs equipped with SSL inlets, Restek
Supplier: Avantor
Certificates
About this item
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
- Consistent quality with exceptional batch-to-batch uniformity
- Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.
Frequently Bought Together