Single Taper Inlet Liner for Shimadzu 14A GCs equipped with split/splitless inlets, Restek
Catalogue # RK20861
Fournisseur: Restek
Spécifications
- Description:Split
- Cat. no.:RK20861
- For Use With:Shimadzu® GC
- Supplier No.:20861
Spécifications
À propos de cet article
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
- Consistent quality with exceptional batch-to-batch uniformity
- Low to zero bleed or background contamination
- Internal diameter of 3.5 mm
Superior coverage is delivered even with highly active compounds.