ContactUsLinkComponent
Cyclo Uniliner® Inlet Liner with Top Taper for PerkinElmer Auto SYS™ GCs equipped with split/splitless inlet, Restek
Catalogue # RK20840
Fournisseur:  Restek
undefined
Cyclo Uniliner® Inlet Liner with Top Taper for PerkinElmer Auto SYS™ GCs equipped with split/splitless inlet, Restek
Catalogue # RK20840
Fournisseur:  Restek
Supplier Number:  20840
Spécifications
  • Description:
    Auto SYS Cyclo-Uniliner
  • Cat. no.:
    RK20840
  • For Use With:
    PerkinElmer® GC, Auto SYS GC
  • Supplier No.:
    20840
Spécifications
À propos de cet article
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
  • Consistent quality with exceptional batch-to-batch uniformity
  • Low to zero bleed or background contamination
  • Internal diameter of 4 mm
Superior coverage is delivered even with highly active compounds.