Drilled Cyclo Uniliner® Inlet Liner with Hole near Top and Top Taper for Agilent GCs Equipped with Split/Splitless Inlets, Restek
: Restek
Use of these liners helps ensure an inert GC flow path for higher sensitivity, accuracy, and reproducibility.
- Consistent quality with exceptional batch-to-batch uniformity
- Low to zero bleed or background contamination
Superior coverage is delivered even with highly active compounds.
Frequently Bought Together