ContactUsLinkComponent
¹/₁₆" Capillary Inlet Adapter Fitting Kit, Split/Splitless Fitting for Capillary Columns, ¹/₁₆", Restek
Catalogue # 10856-160
Fournisseur:  Restek
undefined
¹/₁₆" Capillary Inlet Adapter Fitting Kit, Split/Splitless Fitting for Capillary Columns, ¹/₁₆", Restek
Catalogue # 10856-160
Fournisseur:  Restek
Supplier Number:  27184
Spécifications
  • Description:
    1/16" Capillary Inlet Adaptor Fitting Kit
  • Cat. no.:
    10856-160
  • Supplier No.:
    27184
Spécifications
À propos de cet article
Capillary columns are easier to install due to the nut protruding farther from the insulated injection port chamber.
  • 1/16" split/splitless fitting that accepts standard, two-hole capillary ferrules
  • Same column insertion depth as the original manufacturer's equipment
For use with Agilent split/splitless fittings for capillary columns.
Kit includes adapter fitting, 1/16" capillary nut, gold-plated 1.2 mm ID dual Vespel ring inlet seal, one 0.4 mm ID two-hole ferrule, and one 0.5 mm ID two-hole ferrule.