Capillary Installation Gauge for Shimadzu 17A, 2010, and 2014 GCs, Restek
Catalog # RK22333
Supplier: Restek
Specifications
- Description:Capillary Installation Gauge for use with Shimadzu® 17A GC, 2010 GC, and 2014 GC
- Cat. no.:RK22333
- Supplier No.:22333
Specifications
About this item
Use a capillary installation gauge for Shimadzu 17A, 2010, and 2014 GCs to seat ferrules onto a column for consistent installations.
- For use with graphite ferrules only
Ensure reliable, consistent ferrule installation with these capillary installation gauges.
Ordering information: For use with graphite ferrules only.