ContactUsLinkComponent
Click-On In-Line Super Clean® Double Connector, Restek
Click-On In-Line Super Clean® Double Connector, Restek
Catalog # 10855-706
Supplier:  Restek
undefined
Click-On In-Line Super Clean® Double Connector, Restek
Catalog # 10855-706
Supplier:  Restek
Supplier Number:  22479
Specifications
  • Description:
    Click-On In-line Super Clean Double Connector, Stainless Steel
  • Cat. no.:
    10855-706
  • Supplier No.:
    22479
Specifications
About this item
Double connectors are used to effectively connect any click-on trap to a click-on indicator.
  • Stainless steel construction
Each double connector is 3" (8 cm) in length.