ContactUsLinkComponent
Click-On In-Line Super Clean® Indicator, Restek
Catalogue # 10855-012
Fournisseur:  Restek
undefined
Click-On In-Line Super Clean® Indicator, Restek
Catalogue # 10855-012
Fournisseur:  Restek
Supplier Number:  22474
Spécifications
  • Description:
    Click-On In-line Super Clean Indicator (oxygen, moisture)
  • Cat. no.:
    10855-012
  • Supplier No.:
    22474
Spécifications
À propos de cet article
Install an indicator after the Click-On in-line gas filter so there is no confusion about when to replace the traps.
  • Colored indicator for detecting buildup of contaminants
  • Oxygen: green to gray
  • Moisture: beige to clear
Ordering information: Note: Fittings sold separately.