Valco® Adapter Ferrules, ¹/₁₆", Restek
Supplier: Restek
Jump to:
À propos de cet article
Fused silica adapter ferrules are made from Valcon polyimide, a unique, graphite-reinforced composite specially prepared to maximize mechanical stability at temperatures up to 350°C.
- Fabricated from Valcon polyimide
- Stable up to 350°C
The determining factor for selecting adapter ferrule size is the fused silica tubing OD.
4 Options Available Below
Product Details & Documents
Fused silica adapter ferrules are made from Valcon polyimide, a unique, graphite-reinforced composite specially prepared to maximize mechanical stability at temperatures up to 350°C.
- Fabricated from Valcon polyimide
- Stable up to 350°C
The determining factor for selecting adapter ferrule size is the fused silica tubing OD.
Product Family Options
Informations sur le produit
- DésignationMatièreTailleFits Tubing O.D.AvailabilityPrice
- Supplier Number: 20142Spécifications:
More
Specifications
Cat. No.10852-928 - Supplier Number: 20145Spécifications:
More
Specifications
Cat. No.10852-930 - Supplier Number: 20143Spécifications:
More
Specifications
Cat. No.10852-932 - Supplier Number: 20144Spécifications:
More
Specifications
Cat. No.10852-934