Scoring Wafer with Handle, Restek
Scoring Wafer with Handle
97040-464
: Restek
: 23015
Ceramic scoring wafer is straight-edged for cutting fused silica tubing cleanly.
- Unique, ergonomic handle is made of soft, comfortable rubber
- Désignation:Scoring Wafer with Handle