Rethreading Tool, Restek
Supplier:   Restek
Rethreading Tool, Restek
Supplier:  Restek
Certificates
Rethreading Tool, Restek
Rethreading Tool, Restek
2 Options Available Below
Product Details & Documents
Rethreading tools repair worn or damaged threads that come about as a result of consistent use and temperature changes.
  • Multiple uses (fittings, injectors, detectors, etc.)
  • Built-in guide to prevent cross-threading
Repeated installation, removal, and temperature changes cause wear and damage to threads on GC parts. This can cause a poor seal, and oxygen can enter the system, compromising analytical results and possibly destroying expensive analytical columns.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Rethreading Tool
    Discontinued
    Supplier Number: -
  • Rethreading Tool
    Discontinued
    Supplier Number: -
  • Rethreading Tool
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $197.97
    $197.97
    Prix promotionnel pour les nouveaux clients
    Each
       $197.97
    Supplier Number: 23016
  • Rethreading Tool
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $295.85
    $295.85
    Prix promotionnel pour les nouveaux clients
    Each
       $295.85
    Supplier Number: 23017