MXT® 'Y'-Union Connector Kits for Connecting Metal and/or Fused Silica GC Columns, Restek
: Restek
Connector kits for metal or fused silica columns are ideal for connecting guard columns or transfer lines to analytical columns.
- Low dead volume, leak-tight connection
- Reusable
- Siltek® treatment ensures maximum inertness
- Ideal for connecting a guard column or transfer line to an analytical column
- Use to oven temperatures of 350°C
- Available in union and “Y” configurations
- Can also be used for connecting fused silica to metal
Each kit contains the MXT® union; three 1/32-inch nuts; and three, one-piece, fused silica adapters.
Frequently Bought Together