Dual Column mini-Lam Direct Injection Tee Kit for ¹/₄" packed column inlets, Restek
Catalog # 75778-664
Supplier: Avantor
Specifications
- Description:Mini-Lam DI Tee Kit
- Cat. no.:75778-664
- Includes:All fittings and ferrules
- Supplier No.:20436
Specifications
About this item
Inverted cup design offers complete sample vaporization.
- Permits larger sample volumes
- Incorporates a Press-Tight® taper in each outlet leg, to make a perfect, dead-volume-free connection to each analytical column (ODs ranging from 0.4 to 0.8 mm)
- Allows visual confirmation of the column connection
Open-top design makes it easy to pack with glass wool, keeping sample residue from contaminating the cup.