Tubing Burring and Reaming Tool, Restek
Fournisseur: Restek
Certificats
À propos de cet article
Removes burrs and reams tubing.
- Available in 1/16" and 1/8" sizes
Informations sur les accessoires: Inserts are for use with 1/16" tubing dressing tools.
Frequently Bought Together