Protocol Station, Restek | Avantor
Protocol Station, Restek
Supplier:   Restek
Protocol Station, Restek
Supplier:  Restek
Certificates
Protocol Station, Restek
Protocol Station, Restek
4 Options Available Below
Product Details & Documents
The protocol station is designed for convenient wall mounting of high-purity gas regulators.
  • Fabricated in stainless steel or chrome-plated brass
  • Provides ease of use for high-purity gas regulators
Wall mounting provides ease of use, prevents gas regulator damage, and improves safety. Either chrome-plated brass or 316 stainless steel option is complete with a 3-foot, flexible, all-stainless-steel pigtail with armor casing. The CGA connection on the pigtail has an integral check valve in the gland to prevent contamination during cylinder changeout.
Ordering information: Pressure regulator not included. Order separately.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Includes
    Availability
    Price
  • Protocol Station*
    Stainless steel
    CGA 580 (N2, He, Ar)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,540.80
    $1,540.80
    Prix promotionnel pour les nouveaux clients
    Each
       $1,540.80
    Supplier Number: 21327
  • Protocol Station*
    Brass, chrome plated
    CGA 580 (N2, He, Ar)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,387.62
    $1,387.62
    Prix promotionnel pour les nouveaux clients
    Each
       $1,387.62
    Supplier Number: 21347
  • Protocol Station*
    Brass, chrome plated
    CGA 350 (H2, P5)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,373.98
    $1,373.98
    Prix promotionnel pour les nouveaux clients
    Each
       $1,373.98
    Supplier Number: 21348
  • Protocol Station*
    Brass, chrome plated
    CGA 590 (Air)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,406.38
    $1,406.38
    Prix promotionnel pour les nouveaux clients
    Each
       $1,406.38
    Supplier Number: 21349