MSD Conversion Fitting, Restek
MSD Conversion Fitting
Catalog # RK21314
Supplier: Restek
Fournisseur #: 21314
Jump to:
Product Details & Documents
A flat, soft aluminum sealing ring deforms and butt-seals against the MSD interface.
- A standard Vespel® ferrule seals the column and 1/16" stainless steel nut
- Fitting is constructed of nickel-plated brass for longevity and softness
- Use any standard Vespel® or Vespel®/graphite 1/16" ferrule
- Improved design reduces chance of leaks
Ordering information: Includes a 1/16" stainless steel nut and two replacement sealing rings. Order ferrules separately.
Spécifications
- Désignation:MSD Conversion Fitting