Inserts for 4.0 ml WISP 48 Screw-Thread Step Vials, Restek | Avantor
Inserts for 4.0 ml WISP 48 Screw-Thread Step Vials, Restek
Supplier:   Restek
Inserts for 4.0 ml WISP 48 Screw-Thread Step Vials, Restek
Supplier:  Restek
Certificates
Inserts for 4.0 ml WISP 48 Screw-Thread Step Vials, Restek
Inserts for 4.0 ml WISP 48 Screw-Thread Step Vials, Restek
2 Options Available Below
Product Details & Documents
A variety of inserts compatible with screw-thread and crimp-top vials are available in a flat-bottomed format.
  • Screw-thread and crimp-top vial compatibility
  • Can also be used with snap seal vials
  • Polypropylene construction
Inserts for screw-thread, crimp-top, and snap seal vials are made of rugged polypropylene and feature a flat-bottomed design.
Ordering information: Polypropylene inserts available on request (1000-packs only). Also fit 4.0 ml Snap Seal vials.
Product Family Options
Informations sur le produit
  • Désignation
    Capacité
    Bottom Style
    Matière
    Avec bouchon
    Availability
    Price
  • Insert for WISP 48 vials
    0.50 ml
    Flat bottom
    Glass
    Non
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - $59.22
    $59.22
    Prix promotionnel pour les nouveaux clients
    Pack of 100
       $59.22
    Supplier Number: 21787
  • Insert for WISP 48 vials
    0.50 ml
    Flat bottom
    Glass
    Non
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 1,000
    Pack of 1,000 - $392.96
    $392.96
    Prix promotionnel pour les nouveaux clients
    Pack of 1,000
       $392.96
    Supplier Number: 21788