WISP 48 Screw-Thread Step Vials, Restek
WISP 48 Screw-Thread Step Vials, Restek
Supplier:   Restek
WISP 48 Screw-Thread Step Vials, Restek
Supplier:  Restek
Certificates
WISP 48 Screw-Thread Step Vials, Restek
WISP 48 Screw-Thread Step Vials, Restek
4 Options Available Below
Product Details & Documents
WISP 48 screw-thread step vials feature a screw-thread design and an extra-wide orifice size of 13 mm.
  • Feature white graduated markings
  • Clear or amber color
  • Screw-thread format
Vials have a 4 ml capacity.
Product Family Options
Product Information
  • Désignation
    Capacité
    Diameter ×H
    Color
    Avec bouchon
    Availability
    Price
  • WISP 48 Step Vial with White Graduated Marking Spot
    4.00 ml
    15×45 mm
    Clear
    Non
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $119.68
    $119.68
    New Customer Promotional Price
    Pack of 100
       $119.68
    Supplier Number: 24654
  • WISP 48 Step Vial with White Graduated Marking Spot
    4.00 ml
    15×45 mm
    Amber
    Non
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $131.37
    $131.37
    New Customer Promotional Price
    Pack of 100
       $131.37
    Supplier Number: 24656
  • WISP 48 Step Vial with White Graduated Marking Spot
    4.00 ml
    15×45 mm
    Clear
    Non
    Log in for availability
    Log in for availability
    Pack of 1,000
    Pack of 1,000 - $615.15
    $615.15
    New Customer Promotional Price
    Pack of 1,000
       $615.15
    Supplier Number: 24655
  • WISP 48 Step Vial with White Graduated Marking Spot
    4.00 ml
    15×45 mm
    Amber
    Non
    Log in for availability
    Log in for availability
    Pack of 1,000
    Pack of 1,000 - $536.87
    $536.87
    New Customer Promotional Price
    Pack of 1,000
       $536.87
    Supplier Number: 24657