8 mm Screw-Thread Vials with 100 µl Insert, Restek
Supplier: Restek
Jump to:
À propos de cet article
Vials with glass or polypropylene inserts feature 8 mm mouths and screw-thread sealing format.
- Plastic fabrication
- Polypropylene or glass inserts
- 8 mm orifice
- Screw-thread format
Vials have a 100 μl capacity.
Ordering information: Amber plastic vial with clear glass insert available on request. (1000-packs only)
3 Options Available Below
Product Details & Documents
Vials with glass or polypropylene inserts feature 8 mm mouths and screw-thread sealing format.
- Plastic fabrication
- Polypropylene or glass inserts
- 8 mm orifice
- Screw-thread format
Vials have a 100 μl capacity.
Ordering information: Amber plastic vial with clear glass insert available on request. (1000-packs only)
Product Family Options
Informations sur le produit
- DésignationCapacitéMatièreColorAvec bouchonAvailabilityPrice
- Screw-thread vial with 100 µl PP insert2.00 mlPlasticClearNonLog in pour la disponibilitéLog in pour la disponibilitéPack of 100Pack of 100 - $67.42$67.42Prix promotionnel pour les nouveaux clientsPack of 100$67.42Supplier Number: 24623
- Screw-thread vial with 100 µl PP insert2.00 mlPlasticClearNonLog in pour la disponibilitéLog in pour la disponibilitéPack of 1,000Pack of 1,000 - $380.49$380.49Prix promotionnel pour les nouveaux clientsPack of 1,000$380.49Supplier Number: 24624
- Screw-thread vial with 100 µl glass insert2.00 mlPlasticClearNonLog in pour la disponibilitéLog in pour la disponibilitéPack of 100Pack of 100 - $372.59$372.59Prix promotionnel pour les nouveaux clientsPack of 100$372.59Supplier Number: 24625