High-Temperature String, Restek
: Restek
Use high-temperature string to restring capillary columns, attach column connectors to column cages, or hold the column in the GC oven.
Capable of withstanding temperatures to 400 °C.
Use high-temperature string to restring capillary columns, attach column connectors to column cages, or hold the column in the GC oven.
Capable of withstanding temperatures to 400 °C.