Pocket Magnifier, Restek
Catalog # RK20124
Supplier: Restek
Specifications
- Magnification:10×
- Description:Pocket Magnifier
- Cat. no.:RK20124
- Supplier No.:20124
Specifications
About this item
10X magnification makes it easy to see the column end to verify a square cut.
- Small and easy to handle