Staticide® Static Sentry, ACL Staticide
Staticide® Static Sentry, ACL Staticide
Staticide® Static Sentry, Aerosol Can, 340 g (12 oz.)
Catalog # 75780-062
Supplier:   ACL Staticide
Supplier #:  2006
Staticide® Static Sentry, ACL Staticide
Catalog # 75780-062
Supplier:  ACL Staticide
Staticide® Static Sentry, ACL Staticide
Staticide® Static Sentry, ACL Staticide
Staticide® Static Sentry, Aerosol Can, 340 g (12 oz.)
Price
$86.50
Log in pour la disponibilité
Each(1/Itm/Bx) Case of 12 (1/ Itm/Bx)
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents
Long lasting topical anti-static spray is ideal for eliminating charge generation on both porous and non-porous materials
Staticide® Static Sentry can be used on a variey of surfaces, including laminate flooring, carpeting, cabinetry, work surfaces, paper, polycarbonate, acrylic, PET, and glass. Safe to use on plastic and fabrics. No tribocharging to prevent dust attraction.
The spray works instantly, and is ozone safe and humidity independent. Ideal for use in offices, laboratories, institutions, and manufacturing facilities.
Certifications: RoHS compliant. Meets MIL-B-81705 Federal Test Standard 101.
Spécifications
  • Désignation:
    Staticide® Static Sentry
  • Taille:
    340 g (12 oz.)
  • Packaging:
    Aerosol Can