À propos de cet article
Stericheck is a presterilized, nontoxic, non-pyrogenic, and ready to use sterility testing device designed for closed loop sterility testing of Large Volume Parenterals (LVP). The LVP series is specifically designed for use with parenterals in large glass bottles.
- Non-coring needle design
- Pre-installed color coded clamps
- Raised transparent vent
- Customized needle connections
- Validated for equal sample distribution
- Compatible with Steripump or other available sterility testing equipment
The LVP system includes a single vented needle connected to PVC twin-tubing to transfer fluids at high flow rates
ETO Sterilized models can be used under a laminar flow hood. Gamma Sterilized models can be used in isolators or laminar flow hoods. PVDF Membrane models can be used for liquids with inhibitory properties, such as antibiotics. CN Membrane models can be used for all types of pharmaceutical products.
Raised vents prevent air flow obstruction due to fluid logging, and 'L' shaped flow directors prevent frothing. A tough membrane helps withstand inadvertent back pressure. Archivable sterilization indicator included in each pack.