Human Recombinant CTRP5 (from E. coli)
CTRP5 (C1qTNF-related protein 5; C1QTNF5) belongs to a highly conserved family of adiponectin paralogs. CTRP5 mediates activation of AMP-activated protein kinase (AMPK) in muscle and liver cells, thereby regulating glucose and lipid metabolism. Serum levels of CTRP5 are significantly higher in obese/diabetic animal models compared to normal controls. Furthermore, CTRP5 may be a putative biomarker for mitochondrial dysfunction. Defects in C1QTNF5 are a cause of late-onset retinal degeneration (LORD).
- Fusion-Tag: His Tag
This recombinant proteins is for research use only.
- Taille:50 μg
- Cat. No.:75793-764
- Protein/peptide type:Recombinant
- Protein/peptide name:CTRP5
- Storage conditions:Working aliquots are stable for up to 3 months when stored at −20 °C.
- Formulation:Lyophilized from a 0.2um-filtered solution in 55mM TRIS-Cl, pH 8.2, containing 150mM NaCl.
- Protein synonyms:C1QTNF5|Complement C1q Tumor Necrosis Factor-related Protein 5
- Purity:>90% (SDS-PAGE)
- Tag sequence:His Tag
- Sequence:16-243
- Species:Human
- Source:E. coli
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



