Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Mouse Recombinant CD74

Mouse Recombinant CD74

0.05 mg
Catalog # 75791-772
Supplier:   Prosci
Supplier #:  92-605
Mouse Recombinant CD74
Catalog # 75791-772
Supplier:  Prosci
CAS Number:  
Mouse Recombinant CD74

0.05 mg

CAS Number:  
Price
$896.33
Log in pour la disponibilité
Each (50µG)
Minimum order quantity is 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Mouse HLA class II histocompatibility antigen gamma chain (CD74), is a single-pass type II membrane protein that in humans is encoded by the CD74 gene. It contains 1 thyroglobulin type-1 domain. CD74 Plays a critical role in MHC class II antigen processing by stabilizing peptide-free class II alpha/beta heterodimers in a complex soon after their synthesis and directing transport of the complex from the endoplasmic reticulum to compartments where peptide loading of class II takes place.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Spécifications

Spécifications

  • Taille:
    0.05 mg
  • Cat. No.:
    75791-772
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Protein/peptide name:
    CD74
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Protein synonyms:
    CD74 antigen|Cluster of Differentiation 74|HLA class II histocompatibility antigen gamma chain|CD74|HLA-DR antigens-associated invariant chain
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Tag sequence:
    C-6 His tag
  • Sequence:
    Gln56-Leu215
  • Species:
    Mouse
  • Molecular weight:
    19.4 kD

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.