Human Recombinant Sentrin-specific Protease 2 (from E. coli)
0.05 mg
Catalog # 75791-038
Supplier: Prosci
Fournisseur #: 92-222
Human Recombinant Sentrin-specific Protease 2 (from <i>E. coli</i>)
Catalog # 75791-038
Supplier: Prosci
CAS Number:
Jump to:
- L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
- Return Policy
Product Details & Documents
SENP2 is an enzyme that belongs to the peptidase C48 family
This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His, SUMO tag
SENP2 is a protease that catalyzes two essential functions in the SUMO pathway: processing of full-length SUMO1, SUMO2 and SUMO3 to their mature forms and deconjugation of SUMO1, SUMO2 and SUMO3 from targeted proteins. SUMO1 is a small ubiquitin-like protein that can be covalently conjugated to other proteins. It has been implicated as a down-regulator of CTNNB1 levels and may therefore be a modulator of the Wnt pathway.
Spécifications
Spécifications
- Taille:0.05 mg
- Enzyme type:Recombinant
- Enzyme synonyms:Sentrin-specific protease 2, Axam2, SMT3-specific isopeptidase 2, Sentrin/SUMO-specific protease SENP2, KIAA1331, SENP2., SENP2
- Purity:>95% as determined by reducing SDS-PAGE
- Sequence:Asp363-Leu589
- Source:E. coli
- Molecular weight:39.2 kD
- Cat. No.:75791-038
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Storage conditions:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze−thaw cycles.
- Formulation:Supplied as a 0.2 µm filtered solution of 50 mM HEPES,5% glycerol pH 7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH₂O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tag sequence:N-6 His, SUMO tag
- Tested applications:Biological assays
- Species:Human
- Enzyme name:Sentrin-specific Protease 2



