Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Human Recombinant LMCD1 (from E. coli) | Avantor

Human Recombinant LMCD1 (from E. coli)

0.05 mg
Catalog # 75791-014
Supplier:   Prosci
Fournisseur #:  92-210
Human Recombinant LMCD1 (from <i>E. coli</i>)
Catalog # 75791-014
Supplier:  Prosci
CAS Number:  
Human Recombinant LMCD1 (from E. coli)

0.05 mg

CAS Number:  
Price
$1,570.22
Log in pour la disponibilité
Each (50µG)
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

LMCD1 is transcriptional cofactor which contains a cysteine-rich domain in the N-terminal region and 2 LIM domains in the C-terminal region. It also has several potential phosphorylation and N-myristoylation sites and a single potential N-glycosylation site. LMCD1 is expressed in many tissues with highest abundance in skeletal muscle. LMCD1 restricts GATA6 function by inhibiting DNA-binding, resulting in repression of GATA6 transcriptional activation of downstream target genes. It plays a critical role in the development of cardiac hypertrophy via activation of calcineurin/nuclear factor of activated T-cells signaling pathway.

  • Fusion-Tag: N, C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Spécifications

Spécifications

  • Taille:
    0.05 mg
  • Protein/peptide name:
    LMCD1
  • Protein synonyms:
    Dyxin|LMCD1|LIM and cysteine-rich domains protein 1
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Sequence:
    Met1-Ser365
  • Source:
    E. coli
  • Molecular weight:
    44 kD
  • Cat. No.:
    75791-014
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tag sequence:
    N, C-6 His tag
  • Tested applications:
    Bioassay
  • Species:
    Human

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.