Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Human Recombinant UBE2C (from E. coli)

0.05 mg
Catalog # 75790-872
Supplier:   Prosci
 :  92-139
Human Recombinant UBE2C (from <i>E. coli</i>)
Catalog # 75790-872
Supplier:  Prosci
CAS Number:  
Human Recombinant UBE2C (from E. coli)

0.05 mg

CAS Number:  
Price
1 247,54 $
Log in for availability
Each (50µG)
 
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Ubiquitin-Conjugating Enzyme E2 C (UBE2C) is a 179 amino acid enzyme that belongs to the Ubiquitin-Conjugating Enzyme family. UBE2C is highly expressed in tumor tissues and at low levels in most adult normal tissues. UBE2C is required for the destruction of mitotic cyclins and for cell cycle progression. UBE2C accepts Ubiquitin from the E1 complex and catalyzes its covalent attachment to other proteins. It acts as an essential factor of the anaphase promoting complex/cyclosome (APC/C), which has E3 ubiquitin ligase activity, and targets for destruction substrates from the preceding mitosis (Cyclin A, Cyclin B, Securin, Geminin).

  • Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Specifications

Specifications

  • Taille:
    0.05 mg
  • Protein/peptide name:
    UBE2C
  • Protein synonyms:
    UBE2C|UbcH10|Ubiquitin Carrier Protein C|Ubiquitin-Conjugating Enzyme E2 C|Ubiquitin-Protein Ligase C
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Sequence:
    Met1-Pro179
  • Source:
    E. coli
  • Molecular weight:
    23.3 kD
  • Cat. No.:
    75790-872
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Formulation:
    Supplied as a 0.2 um filtered solution of 20mM PB,150mM NaCl, pH 7.0. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tag sequence:
    N-6 His tag
  • Tested applications:
    Bioassay
  • Species:
    Human

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.