Mouse Recombinant Serpin E1
0.05 mg
Catalog # 75790-426
Supplier: Prosci
Supplier #: 91-895
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Plasminogen activator inhibitor-1 (serpin E1) is a serine protease inhibitor which belongs to the serpin family. Serpin E1 acts as 'bait' for tissue plasminogen activator, urokinase, protein C and matriptase-3/TMPRSS7. Its rapid interaction with PLAT may function as a major control point in the regulation of fibrinolysis.
- Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Specifications
Specifications
- Taille:0.05 mg
- Protein/peptide name:Serpin E1
- Protein synonyms:Mr1|Endothelial plasminogen activator inhibitor|Plasminogen activator inhibitor 1|Planh1|Pai1|Serpin E1|PAI-1
- Purity:> 95% as determined by reducing SDS-PAGE
- Sequence:Leu24-Ser402
- Molecular weight:43.7 kD
- Cat. No.:75790-426
- Protein/peptide type:Recombinant
- Endotoxin content:<0.1 ng/ug (1 IEU/ug) as determined by LAL test.
- Storage conditions:Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Formulation:Lyophilized from a 0.2 um filtered solution of 20mM PB,150mM NaCl,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
- Tag sequence:C-6 His tag
- Tested applications:Bioassay
- Species:Mouse



