Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
 

Human Recombinant PRL2 (from E. coli)

0.05 mg
Catalog # 75788-620
Supplier:   Prosci
 :  91-619
Human Recombinant PRL2 (from <i>E. coli</i>)
Catalog # 75788-620
Supplier:  Prosci
CAS Number:  
Human Recombinant PRL2 (from E. coli)

0.05 mg

CAS Number:  
Price
$1,055.50
Log in pour la disponibilité
Each (50µG)
 
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

PTP4A2, also known as PRL2 or PTPCAAX2, is short for Protein tyrosine phosphatase type IVA 2. This protein exists in cell membrane, cytoplasm,endosome and membrane. PTP4A2 is often farnesylated during post-translational modification. Farnesylation is required for membrane targeting and for interaction with RABGGTB. The unfarnesylated forms are redirected to the nucleus and cytosol. It can stimulate progression from G1 into S phase during mitosis and promotes tumors. It also inhibits geranylgeranyl transferase type II activity by blocking the association between RABGGTA and RABGGTB.

  • Fusion-Tag: C-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Spécifications

Spécifications

  • Taille:
    0.05 mg
  • Protein/peptide name:
    PRL2
  • Protein synonyms:
    OV-1|PTP4A2|HU-PP-1|Protein tyrosine phosphatase type IVA 2|PRL-2|PTP(CAAXII)|Protein-tyrosine phosphatase 4a2|Protein-tyrosine phosphatase of regenerating liver 2
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Sequence:
    Met1-Gln167
  • Source:
    E. coli
  • Molecular weight:
    20.2 kD
  • Cat. No.:
    75788-620
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of PBS,pH7.4. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tag sequence:
    C-6 His tag
  • Tested applications:
    Bioassay
  • Species:
    Human

Spécifications