Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Human Recombinant GRP (from E. coli) | Avantor

Human Recombinant GRP (from E. coli)

0.05 mg
Catalog # 75789-136
Supplier:   Prosci
Fournisseur #:  91-210
Human Recombinant GRP (from <i>E. coli</i>)
Catalog # 75789-136
Supplier:  Prosci
CAS Number:  
Human Recombinant GRP (from E. coli)

0.05 mg

CAS Number:  
Price
$1,514.86
Log in pour la disponibilité
Each (50µG)
La quantité minimale de commande est de 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

U6 snRNA-associated Sm-like protein LSm4 (LSM4) is a member of the snRNP Sm proteins family. Sm-like proteins contain the Sm sequence motif and are thought to form a stable heteromer present in tri-snRNP particles, which are important for pre-mRNA splicing. LSM4 forms a heteromer with a donut shape. The complexes are involved in various steps of RNA metabolism. LSM4 binds specifically to the 3-terminal U-tract of U6 snRNA. LSM4 contributes RNA protein interactions and structural changes which are essential during ribosomal subunit assembly.

  • Fusion-Tag: N-6 His tag

This recombinant protein can be used for biological assays. For research use only.

Spécifications

Spécifications

  • Taille:
    0.05 mg
  • Protein/peptide name:
    GRP
  • Protein synonyms:
    LSM4|GRP|Glycine-Rich Protein|U6 snRNA-Associated Sm-like Protein LSm4
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Sequence:
    Met1-Gln139
  • Source:
    E. coli
  • Molecular weight:
    17.5 kD
  • Cat. No.:
    75789-136
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Storage conditions:
    Lyophilized protein should be stored at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Formulation:
    Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl, 100mM NaCl, 1mM DTT, pH 8.0 . Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the Lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tag sequence:
    N-6 His tag
  • Tested applications:
    For Biological assays
  • Species:
    Human

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.