Saisie de commande
Canada
Lien pour commandesContactUsLinkComponent
Human Recombinant UBE2A (from <i>E. coli</i>)

Human Recombinant UBE2A (from E. coli)

0.05 mg
Catalog # 75789-042
Supplier:   Prosci
Supplier #:  91-163
Human Recombinant UBE2A (from <i>E. coli</i>)
Catalog # 75789-042
Supplier:  Prosci
CAS Number:  
Human Recombinant UBE2A (from E. coli)

0.05 mg

CAS Number:  
Price
$666.01
Log in pour la disponibilité
Each (50µG)
Minimum order quantity is 1
  • L'article nécessite un contrôle de la température pour le stockage et la livraison avec des frais supplémentaires. Il n'est pas éligible au retour pour des raisons de sécurité et de qualité. Tenez compte des exigences avant d'acheter.
  • Return Policy
Product Details & Documents

Ubiquitin-Conjugating Enzyme E2 (UBE2A) is a member of the E2 Ubiquitin-Conjugating Enzyme family. The modification of proteins with ubiquitin is an important cellular mechanism for targeting abnormal or short-lived proteins for degradation. Ubiquitination involves at least three classes of enzymes: ubiquitin-activating enzymes, or E1s, ubiquitin-conjugating enzymes, or E2s, and ubiquitin-protein ligases, or E3s. UBE2A catalyzes the covalent attachment of ubiquitin to other proteins. UBE2A is required for postreplication repair of UV-damaged DNA. UBE2A Interacts with RAD18 and WAC.

  • Fusion-Tag: N-GST, 6 His tag

This recombinant protein can be used for biological assays. For research use only.

Spécifications

Spécifications

  • Taille:
    0.05 mg
  • Protein/peptide name:
    UBE2A
  • Protein synonyms:
    Ubiquitin Carrier Protein A|RAD6 Homolog A|Ubiquitin-Conjugating Enzyme E2 A|RAD6A|hHR6A|HR6A|UBE2A|Ubiquitin-Protein Ligase A
  • Purity:
    > 95% as determined by reducing SDS-PAGE
  • Sequence:
    Met1-Cys152
  • Source:
    E. coli
  • Molecular weight:
    49 kD
  • Cat. No.:
    75789-042
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    <0.1 ng/ug (1 IEU/ug) as determined by LAL test.
  • Storage conditions:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Formulation:
    Supplied as a 0.2 um filtered solution of 50mM HEPES, 150mM NaCl, 2mM DTT, 10% Glycerol, pH 7.5. Always centrifuge tubes before opening. Do not mix by vortex or pipetting. It is not recommended to reconstitute to a concentration less than 100 ug/ml. Dissolve the lyophilized protein in ddH2O. Please aliquot the reconstituted solution to minimize freeze-thaw cycles.
  • Tag sequence:
    N-GST, 6 His tag
  • Tested applications:
    Bioassay
  • Species:
    Human

Spécifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.