Cimarec™ Basic Hotplates, Thermo Scientific
Cimarec™ Basic Hotplates, Thermo Scientific
Cimarec™ Basic Hotplates, Thermo Scientific
A sleek, rugged design and construction reduces the chance of top plate breakage and promotes durability, for economical high performance.
  • Reach maximum temperature with in 8 minutes
  • Solid ceramic top plate cleans easily, resists acids and alkalis
  • New top-plate design reduces chance of breakage
  • Spillaway design diverts spills away from internal components
  • Control knob allows precise temperature control
  • Integral ring-stand holder accommodates 1.3 cm (1/2") diameter support rod
Product Family Options
  • Désignation
    Electrical
    Plage de température
    Top Plate Dimensions
    Overall Dimensions
  • Basic Hotplates, Ceramic Top Plate
    120 V, 60 Hz, 3.21 A, 385 W
    150...538 °C (302...1000 °F)
    10.8W×10.8D cm (4¼×4¼")
    20W x 14.5D x 12.7H cm (79/10 x 57/10 x 5")
    Each
    Each - $537.75
    $537.75
    Each
       $537.75
     : HP194515
    Specifications
    Cat. No.
    97027-350
  • Basic Hotplates, Ceramic Top Plate
    120 V, 60 Hz, 8.96 A, 1062 W
    150...538 °C (302...1000 °F)
    18.4W×18.4D cm (7¼×7¼")
    28.2W x 21.1D x 12.7H cm (111/10 x 83/10 x 5")
    Each
    Each - $776.63
    $776.63
    Each
       $776.63
     : HP194825
    Specifications
    Cat. No.
    97027-356