Cimarec™ Basic Stirrers, Thermo Scientific | Avantor
Cimarec™ Basic Stirrers, Thermo Scientific
Cimarec™ Basic Stirrers, Thermo Scientific
Certificates
Cimarec™ Basic Stirrers, Thermo Scientific
1 Options Available Below
Product Details & Documents
Reach stirring speeds from 250 to 2500 rpm with Cimarec™ Basic Stirrers
  • Control knobs allow precise speed setting
  • Top-plate design reduces chance of breakage
  • Spillaway design directs spills away from internal components
  • Integral ring-stand holder accommodates 1.3 cm (1/2") diameter support rod
Ideal for economical, general-purpose use.
Delivery information: Stirrers include TFE-coated stir bar.
Product Family Options
Informations sur le produit
  • Désignation
    Speed Range
    Top Plate Dimensions
    Overall Dimensions
    Electrical
    Availability
    Price
  • Basic Stirrers, Ceramic (Black Friday version)
    250 - 2500 rpm
    18.4×18.4 cm (7¼×7¼")
    28.2W×21.1D×12.7H cm (111/10×83/10 ×5)"
    120 V, 23 W
    Discontinued
    Supplier Number: S194925
    Spécifications:
    New Product
    New Product
  • Basic Stirrers, Ceramic
    250 - 2500 rpm
    10.8×10.8 cm (4¼×4¼")
    20W×14.5D×12.7H cm (79/10×57/10 ×5)"
    120 V, 21 W
    Discontinued
    Supplier Number: S194615
  • Basic Stirrers, Ceramic
    250 - 2500 rpm
    18.4×18.4 cm (7¼×7¼")
    28.2W×21.1D×12.7H cm (111/10×83/10 ×5)"
    120 V, 23 W
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,303.97
    $1,303.97
    Prix promotionnel pour les nouveaux clients
    Each
       $1,303.97
    Supplier Number: S194925