Dualmat™ Static Dissipative Mats, Bulk Roll, Light Blue, ACL
Supplier: ACL Staticide
Certificates
About this item
Dualmat™ is a heavy duty ESD-safe rubber material designed for use on tables and grounded work bench surfaces
These dual layer, bulk roll mats are supplied in 40 foot lengths. They are available in 24", 30", 36", and 48" widths. Dualmats™ are low outgassing and low VOC, making them ideal for use in cleanrooms and other controlled environments. Additionally, they are UV protected so the color will not fade.
Dissipative on the colored side and conductive on the back side, the design makes it ideal for use with constant monitors. Embossed finish reduces glare, Resistance is 10⁶ to 10⁸ ohms. Electricals warranted for the lifetime of the mat.
Certifications: Compliant to ANSI/ESD S20.20 and RoHS.
Frequently Bought Together