Cap Strips for Tubes, Molecular Bio Products
Fournisseur: Thermo Scientific
Certificats
À propos de cet article
Seal tubes tightly for storage
- Made of polyethylene
- For 0.75 and 1.2 mL sterile tubes
- Available pre-sterilized as the material will not withstand autoclaving
Can be used with rows of tubes or cut apart for individual samples.
Frequently Bought Together