PLA Filament
Supplier: PICOSOLUTIONS GROUP INC SE
Jump to:
À propos de cet article
This PLA filament is perfect for most 3D printers.
- Perfect for applications in schools
- Made in the USA
PLA (Poly Lactic Acid) is a popular and reliable type of thermoplastic used in 3D printing. Its relatively low melting temperature, low thermal shrinkage, and low odor makes it perfect for applications in schools.
Diameter: 1.75 mm
Spool size: 1 kg
Print temperature: 210 to 220 °C
Bed temperature: 50 to 70 °C
5 Options Available Below
Product Details & Documents
This PLA filament is perfect for most 3D printers.
- Perfect for applications in schools
- Made in the USA
PLA (Poly Lactic Acid) is a popular and reliable type of thermoplastic used in 3D printing. Its relatively low melting temperature, low thermal shrinkage, and low odor makes it perfect for applications in schools.
Diameter: 1.75 mm
Spool size: 1 kg
Print temperature: 210 to 220 °C
Bed temperature: 50 to 70 °C
Product Family Options
Informations sur le produit
- DésignationColorAvailabilityPrice
- Supplier Number: PS-5001
- Supplier Number: PS-5000
- Supplier Number: PS-5017
- Supplier Number: PS-5016
- Supplier Number: PS-5004
- Discontinued-Supplier Number: -