Somso® Herbaceous Dicot Stem Section Model
Herbaceous Dicot Stem Section, 1, 32 cm, 26 cm, 14 cm
Catalog # 470029-846
Supplier: MARCUS SOMMER
Fournisseur #: BOS 22
Jump to:
Product Details & Documents
Explore Vascular Structure In This Detailed Model.
- Shows Complex Vascular Structure
- Key Printed On Base For Easy Reference
- 5 Year Manufacturers Warranty
Designed specifically to show the development of vascular bundles of dicots. This model will provide you with years of use. Enlarged approx. 550 times, in SOMSO-Plast®. After Prof. Dr. W. Jung. In one piece. On a green base with explanatory note.
Spécifications
- Désignation:Herbaceous Dicot Stem Section
- Hauteur:14 cm
- Longueur:26 cm
- Width:32 cm
- No. of Parts:1