Colorimetric and Fluorogenic Enzyme Substrates, Biotium
Colorimetric and Fluorogenic Enzyme Substrates, Biotium
Supplier:   Biotium
Colorimetric and Fluorogenic Enzyme Substrates, Biotium
Supplier:  Biotium
Certificates
Colorimetric and Fluorogenic Enzyme Substrates, Biotium
17 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Popular chromogenic and fluorogenic substrates for phosphatases, peroxidases, and other enzymes.
  • Wide selection of popular substrates for alkaline phosphatase, horseradish peroxidase, beta-galactosidase, and other common enzymes
  • Chromogenic substrates for absorbance-based detection or light microscopy
  • Fluorogenic substrates for highly sensitive fluorescence-based ELISA
  • Substrate sampler kits available
Biotium offers a number of popular substrates for alkaline phosphatase, beta-galactosidase, and horseradish peroxidase which are widely used for chromogenic or fluorogenic ELISA, immunohistochemistry, or blotting applications.
Beta-glucuronidase substrates are widely used to detect E. coli bacterial contamination in food and water, and also in reporter gene assays in plants.
X-beta-D-cellobioside is a substrate for beta-cellobiosidase.
ABTS is a registered trademark of Roche. Amplex Red is a registered trademark of Thermo Fisher Scientific.
Product Family Options
Chromogenic Phosphatase Substrates
Product Information
  • Désignation
    Taille
    Availability
    Price
  • BCIP/NBT Kit
    1 kit
    Log in for availability
    Log in for availability
    Each
    Each - $313.28
    $313.28
    Each
       $313.28
    Supplier Number: 10003
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • BCIP Red
    100 mg
    Log in for availability
    Log in for availability
    Each (100mg)
    Each (100mg) - $314.94
    $314.94
    Each (100mg)
       $314.94
    Supplier Number: 10004
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • BCIP Red/NBT Kit
    1 kit
    Log in for availability
    Log in for availability
    Each
    Each - $400.14
    $400.14
    Each
       $400.14
    Supplier Number: 10005
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • BCIP Pink
    100 mg
    Log in for availability
    Log in for availability
    Each (100mg)
    Each (100mg) - $371.19
    $371.19
    Each (100mg)
       $371.19
    Supplier Number: 10006
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • BCIP Pink/NBT Kit
    1 kit
    Log in for availability
    Log in for availability
    Each
    Each - $435.68
    $435.68
    Each
       $435.68
    Supplier Number: 10007
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Chromogenic Phosphatase Substrate Sampler Kit
    1 kit
    Log in for availability
    Log in for availability
    Each
    Each - $442.19
    $442.19
    Each
       $442.19
    Supplier Number: 10022
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Fluorogenic Phosphatase Substrates
Product Information
  • Désignation
    Taille
    Availability
    Price
  • FDP (high purity)
    5 mg
    Log in for availability
    Log in for availability
    Each (5mg)
    Each (5mg) - $433.88
    $433.88
    Each (5mg)
       $433.88
    Minimum order quantity is 1
    Supplier Number: 10030
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Chromogenic Beta-Galactosidase Substrates
Product Information
  • Désignation
    Taille
    Availability
    Price
  • Red-Beta-D-Gal
    100 mg
    Log in for availability
    Log in for availability
    Each (100mg)
    Each (100mg) - $271.47
    $271.47
    Each (100mg)
       $271.47
    Supplier Number: 10012
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Rose-Beta-D-Gal
    100 mg
    Log in for availability
    Log in for availability
    Each (100mg)
    Each (100mg) - $271.47
    $271.47
    Each (100mg)
       $271.47
    Supplier Number: 10013
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Purple-Beta-D-Gal
    25 mg
    Log in for availability
    Log in for availability
    Each (25mg)
    Each (25mg) - $271.47
    $271.47
    Each (25mg)
       $271.47
    Supplier Number: 10014
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Green-Beta-D-Gal
    25 mg
    Log in for availability
    Log in for availability
    Each (25mg)
    Each (25mg) - $305.14
    $305.14
    Each (25mg)
       $305.14
    Supplier Number: 10015
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Beta-Galactosidase Substrate Sampler Kit
    1 kit
    Log in for availability
    Log in for availability
    Each
    Each - $424.24
    $424.24
    Each
       $424.24
    Supplier Number: 10024
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Chromogenic Beta-Glucuronidase Substrates
Product Information
  • Désignation
    Taille
    Availability
    Price
  • X-GlcU, Na (X-GlcA)
    25 mg
    Log in for availability
    Log in for availability
    Each (25mg)
    Each (25mg) - $287.05
    $287.05
    Each (25mg)
       $287.05
    Supplier Number: 10017
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Red-Beta-D-GlcU, CHA (Magenta-Beta-D-GlcUA)
    25 mg
    Log in for availability
    Log in for availability
    Each (25mg)
    Each (25mg) - $298.62
    $298.62
    Each (25mg)
       $298.62
    Supplier Number: 10018
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Rose-Beta-D-GlcU, CHA (Salmon-Beta-D-GlcUA)
    25 mg
    Log in for availability
    Log in for availability
    Each (25mg)
    Each (25mg) - $298.62
    $298.62
    Each (25mg)
       $298.62
    Supplier Number: 10019
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Beta-Glucuronidase Substrate Sampler Kit
    1 kit
    Log in for availability
    Log in for availability
    Each
    Each - $424.24
    $424.24
    Each
       $424.24
    Supplier Number: 10025
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Fluorogenic Beta-Gluocuronidase Substrates
Product Information
  • Désignation
    Taille
    Availability
    Price
  • MUG (MUGlcU; 4-MU-glcUA)
    100 mg
    Log in for availability
    Log in for availability
    Each (100mg)
    Each (100mg) - $208.33
    $208.33
    Each (100mg)
       $208.33
    Supplier Number: 10020
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.