The IN6430 Ionizing Air Gun is an effective static eliminating device that integrates best-in-class AC performance, mechanical, and ergonomic design features into a superior and affordable package
The IN6430 uses Ptec™, a specialized piezoceramic technology, to create a high frequency AC ionizing output with exceptional balance and decay performance. This device eliminates high voltage cables by locating the power supply in the ionizing gun handle. An OSHA compliant emitter nozzle protects both the operator and ionizer. Emitter contamination and wear are virtually non-existent with AC ionizing guns.
The high quality air filters ensure that the air flowing through the IN6340 is clean and free from contaminates. The Micro Filter features a hollow fiber membrane that provides excellent filtration and has a long service life. The compact design is easy to install and all materials used are compatible for the most stringent applications.
Typical AC ionizing guns require stiff and bulky high voltage (HV) cables to connect the gun to the power supply. Over time, these HV cables fail. DC ionizing guns eliminate HV cables, but ion balance is limited. DC emitter points are vulnerable to physical damage, contamination, and erosion, all during normal use.
- Catalog No:
- 10837-132
- 10837-098
- 10837-130
- 10837-094
- Decay time:
- 1000 - 100 V, <2 s at 12"
- 1000 - 100 V, <2 s at 12"