Ionizing Nozzles, Transforming Technologies
À propos de cet article
Transforming Technologies’ Ionizing Nozzles can handle unique and difficult point-of-use ionization applications with interchangeable nozzle tips
Ionizing Nozzles are compatible with all IN Gun series ionizers.
Ionizing nozzles can be mounted in hard to reach areas that larger ionizers simply cannot handle. Nozzle tips are designed to spread ionized air in many unique ways. Wide angle, long range, bendable, luer lock, or manifold tips may be substituted for the N0010 tip, depending upon the application. (N0010 is the standard tip sold with Transforming Technologies Ionizing Nozzles).
8 Options Available Below
Product Details & Documents
Transforming Technologies’ Ionizing Nozzles can handle unique and difficult point-of-use ionization applications with interchangeable nozzle tips
Ionizing Nozzles are compatible with all IN Gun series ionizers.
Ionizing nozzles can be mounted in hard to reach areas that larger ionizers simply cannot handle. Nozzle tips are designed to spread ionized air in many unique ways. Wide angle, long range, bendable, luer lock, or manifold tips may be substituted for the N0010 tip, depending upon the application. (N0010 is the standard tip sold with Transforming Technologies Ionizing Nozzles).
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: N0010Spécifications:
More
Specifications
Cat. No.10837-134 - Supplier Number: N0020Spécifications:
More
Specifications
Cat. No.10837-062 - Supplier Number: N0030Spécifications:
More
Specifications
Cat. No.10837-064 - Supplier Number: N0040Spécifications:
More
Specifications
Cat. No.10837-066 - Supplier Number: N0050-8Spécifications:
More
Specifications
Cat. No.10837-074 - Supplier Number: N0050-12Spécifications:
More
Specifications
Cat. No.10837-068 - Supplier Number: N0050-16Spécifications:
More
Specifications
Cat. No.10837-070 - Supplier Number: N0050-20Spécifications:
More
Specifications
Cat. No.10837-072 - Manifold long (12 - 20") nozzle tipDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.10837-076 - Manifold short (4 - 12") nozzle tipDiscontinued-Supplier Number: -Spécifications:
More
Specifications
Cat. No.10837-078