Toggle Press for Filter Vials
Toggle press for filter vials, metal, non sterile
Catalog # 76808-504
Supplier: THOMSON INSTRUMENT CO
Fournisseur #: 35005
Jump to:
Product Details & Documents
The Thomson filter vial toggle press quickly depresses autosampler-ready filter vials.
Press up to 5 autosampler ready filter vials
- Small footprint sits on bench top
- Works with all Thomson filter vials
Allows for consistency and ergonomic concerns.
Spécifications
- Désignation:Toggle press for filter vials, metal, non sterile