Dehydrant Blends, Epredia
Dehydrant Blends, Epredia
Supplier:   Thermo Scientific
Dehydrant Blends, Epredia
Supplier:  Thermo Scientific
Certificates
Dehydrant Blends, Epredia
Dehydrant Blends, Epredia
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Process and stain histological and cytological specimens with these blends of denatured ethyl alcohol reagent.
  • Blend of ethyl, methyl and isopropyl alcohols
  • Can be used as a dye or stain solvent
  • Miscible with all clearing agents
Process and stain histological and cytological specimens with Epredia™ Dehydrant Blends. These denatured ethyl alcohol reagent blends are specifically designed for processing and staining histological and cytological specimens.
Product Family Options
Informations sur le produit
  • Désignation
    Type de conditionnement
    Taille
    Availability
    Price
  • Dehydrant 70%
    Poly Bottle
    1 gal.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4 (950ml)
    Case of 4 (950ml) - $463.87
    $463.87
    Case of 4 (950ml)
       $463.87
    Supplier Number: 6501
  • Dehydrant 80%
    Poly Bottle
    4 x 1 gal.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4 (0.25 .US Gal)
    Case of 4 (0.25 .US Gal) - $462.04
    $462.04
    Case of 4 (0.25 .US Gal)
       $462.04
    Supplier Number: 6401
  • Dehydrant 95%
    Poly Bottle
    1 gal.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4 (0.25 .US Gal)
    Case of 4 (0.25 .US Gal) - $502.33
    $502.33
    Case of 4 (0.25 .US Gal)
       $502.33
    Supplier Number: 6301
  • Dehydrant 95%
    Metal Drum
    55 gal.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (55US Gal)
    Each (55US Gal) - $4,933.86
    $4,933.86
    Each (55US Gal)
       $4,933.86
    Supplier Number: 6355
  • Dehydrant 100%
    Poly Bottle
    1 gal.
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4 (950ml)
    Case of 4 (950ml) - $463.10
    $463.10
    Case of 4 (950ml)
       $463.10
    Supplier Number: 6201